Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLC1
Entrez Gene ID
10395
Gene Name
DLC1 Rho GTPase activating protein
Gene Aliases
ARHGAP7, HP, STARD12, p122-RhoGAP
Chromosome Location
Chr.8: 13083361 - 13604606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164271.1 NP_001157743.1 8 - 9 2283 138
NM_001316668.1 NP_001303597.1 8 - 9 2471 138
NM_006094.4 NP_006085.2 8 - 9 2541 138
NM_182643.2 NP_872584.2 12 - 13 3976 138
XM_005273374.1 XP_005273431.1 12 - 13 3857 138
XM_017012951.1 XP_016868440.1 12 - 13 3727 138
XM_017012952.1 XP_016868441.1 8 - 9 2274 138
GenBank mRNA AB051510.1 - 12 - 13 3862 138
AF026219.1 - 8 - 9 2494 138
AF035119.1 - 8 - 9 2545 138
AK025544.1 - 8 - 9 2471 138
AK025733.1 - 3 - 4 717 138
AK289734.1 - 8 - 9 2464 138
AK299049.1 - 8 - 9 2283 138
BC054511.1 - 12 - 13 3731 138

View Full Product Details