The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021965.3 | NP_068800.2 | 7 - 8 | 1386 | 78 |
XM_017014808.1 | XP_016870297.1 | 7 - 8 | 1278 | 78 | |
GenBank mRNA | AK297108.1 | - | 7 - 8 | 1208 | 78 |
AL710966.1 | - | 2 - 3 | 190 | 78 |