The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001312653.1 | NP_001299582.1 | 1 - 1 | 459 | 74 |
GenBank mRNA | AK292062.1 | - | 1 - 1 | 456 | 74 |