Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNA22P
Entrez Gene ID
3453
Gene Name
interferon alpha 22, pseudogene
Gene Aliases
IFNAP22
Chromosome Location
Chr.9: 21277688 - 21278563 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036676.1 1 - 1 707 131

View Full Product Details