The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183375.2 | NP_899231.2 | 2 - 3 | 213 | 78 |
XM_011531921.2 | XP_011530223.1 | 1 - 2 | 659 | 78 | |
GenBank mRNA | BC121109.1 | - | 2 - 3 | 217 | 78 |