Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5KL1
Entrez Gene ID
138429
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
PIPKH, bA203J24.5
Chromosome Location
Chr.9: 127920886 - 127931958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135219.1 NP_001128691.1 5 - 6 599 85
XM_017014279.1 XP_016869768.1 5 - 6 1760 85
GenBank mRNA BE881330.1 - 2 - 3 182 85

View Full Product Details