Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSTM2A
Entrez Gene ID
222008
Gene Name
V-set and transmembrane domain containing 2A
Gene Aliases
VSTM2
Chromosome Location
Chr.7: 54542064 - 54571080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182546.3 NP_872352.2 4 - 5 1037 67
GenBank mRNA BI752069.1 - 3 - 4 453 67
BI754879.1 - 4 - 5 578 67

View Full Product Details