The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181501.1 | NP_852478.1 | 25 - 26 | 3232 | 151 |
GenBank mRNA | AK300423.1 | - | 25 - 26 | 3397 | 151 |
BC137121.1 | - | 25 - 26 | 3231 | 151 | |
BC137122.1 | - | 25 - 26 | 3231 | 151 | |
BX640981.1 | - | 14 - 15 | 5855 | 151 | |
BX648284.1 | - | 3 - 4 | 3060 | 151 | |
X68742.1 | - | 24 - 25 | 3096 | 151 |