The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181501.1 | NP_852478.1 | 28 - 29 | 3548 | 145 |
GenBank mRNA | BC137121.1 | - | 28 - 29 | 3547 | 145 |
BC137122.1 | - | 28 - 29 | 3547 | 145 | |
BX648284.1 | - | 6 - 7 | 3376 | 145 |