Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC01315
Entrez Gene ID
102723775
Gene Name
long intergenic non-protein coding RNA 1315
Gene Aliases
lnc-C22orf32-1
Chromosome Location
Chr.22: 42364400 - 42369208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_120595.1 1 - 2 496 67
GenBank mRNA BC038245.1 - 1 - 2 462 67

View Full Product Details