Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
81793
Gene Name
toll like receptor 10
Gene Aliases
CD290
Chromosome Location
Chr.4: 38772238 - 38782990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195106.1 NP_001182035.1 1 - 2 70 121
NM_001195108.1 NP_001182037.1 1 - 2 70 121
NM_030956.3 NP_112218.2 1 - 2 70 121
GenBank mRNA AW504309.1 - 1 - 2 45 121
BP383038.1 - 1 - 2 69 121
DA213264.1 - 1 - 2 66 121
DB226380.1 - 1 - 2 70 121

View Full Product Details