Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HOXC5
- Entrez Gene ID
-
3222
- Gene Name
- homeobox C5
- Gene Aliases
-
CP11, HOX3, HOX3D
- Chromosome Location
-
Chr.12: 54016852 - 54035361 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003084.2 |
|
1 - 2 |
557 |
125 |
GenBank mRNA |
BC034922.1 |
- |
1 - 2 |
299 |
125 |
BC044635.1 |
- |
1 - 2 |
299 |
125 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE394964.1 |
- |
1 - 2 |
277 |
|
Chr. 12 - 54017130 - 54034641 [+] on Build GRCh38 |
125 |
GenBankmRNA |
BE396026.1 |
- |
1 - 2 |
280 |
|
Chr. 12 - 54017129 - 54034649 [+] on Build GRCh38 |
125 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 615 |
MIR615 |
MIRN615, hsa-mir-615, mir-615 |
|
Sense |
Chr. - 54033950 - 54034045 on Build GRCh38 |
Downstream |
uncharacterized LOC440101 |
FLJ12825 |
|
22.9kb
|
Sense |
Chr. - 54058254 - 54122234 on Build GRCh38 |
View Full Product Details