Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
Chromosome Location
Chr.11: 111383828 - 111512388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC022056.1 - 3 - 4 353 127

View Full Product Details