The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001070.4 | NP_001061.2 | 1 - 2 | 444 | 179 |
GenBank mRNA | DB027894.1 | - | 1 - 2 | 395 | 179 |
DB028144.1 | - | 1 - 2 | 444 | 179 |