Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 4 - 4 580 90
NM_001178076.1 NP_001171547.1 2 - 2 346 90
NM_001178077.1 NP_001171548.1 2 - 2 279 90
NM_001673.4 NP_001664.3 4 - 4 654 90
NM_133436.3 NP_597680.2 4 - 4 909 90
NM_183356.3 NP_899199.2 5 - 5 932 90
GenBank mRNA AK302189.1 - 4 - 4 474 90
AK302242.1 - 2 - 2 227 90
AK316224.1 - 2 - 2 281 90
BC008723.2 - 5 - 5 759 90
BC014621.2 - 4 - 4 530 90
BG718826.1 - 4 - 4 624 90
BT007113.1 - 2 - 2 403 90
M15798.1 - 3 - 3 450 90
M27396.1 - 4 - 4 581 90

View Full Product Details