Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
118490
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr.10: 73423579 - 73433561 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR936824.1 - 6 - 6 3818 150

View Full Product Details