The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026575.1 | 1 - 1 | 239 | 130 | |
GenBank mRNA | AK292282.1 | - | 1 - 1 | 239 | 130 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 192 | TMEM192 | -164.9kb | Sense | Chr. - 165076078 - 165112913 on Build GRCh38 | |
Downstream | methylsterol monooxygenase 1 | MSMO1 | DESP4, ERG25, MCCPD, SC4MOL | 47.6kb | Anti-sense | Chr. - 165327666 - 165343164 on Build GRCh38 |