Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
OPA3 | NM_001017989.2 | NP_001017989.2 | ||
NM_025136.3 | 2 | 1063 | NP_079412.1 | |
XM_006723403.3 | 3 | 1025 | XP_006723466.1 | |
AK025840.1 | 2 | 1018 | BAB15255.1 | |
AK098798.1 | BAC05415.1 | |||
AK296429.1 | 2 | 939 | ||
BC005059.1 | 2 | 984 | AAH05059.1 | |
BC047316.1 | AAH47316.1 | |||
BC064146.1 | AAH64146.1 | |||
DB034102.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833848 | Chr19:45457372 - 45644939 on Build GRCh38 | Loss | VASP MIR330 PPM1N RTN2 ERCC1 EML2-AS1 OPA3 EML2 FOSB GPR4 |