Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LILRB1
- Entrez Gene ID
-
10859
- Gene Name
- leukocyte immunoglobulin like receptor B1
- Gene Aliases
-
CD85J, ILT-2, ILT2, LIR-1, LIR1, MIR-7, MIR7, PIR-B, PIRB
- Chromosome Location
-
Chr.19: 54616930 - 54637925 on Build GRCh38
View Full Product Details