The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080671.3 | NP_542402.3 | 2 - 2 | 1210 | 139 |
GenBank mRNA | AY065987.1 | - | 2 - 2 | 924 | 139 |
BC014429.1 | - | 2 - 2 | 931 | 139 |