The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052952.2 | NP_443184.1 | 2 - 2 | 548 | 89 |
XM_017003281.1 | XP_016858770.1 | 2 - 2 | 369 | 89 | |
GenBank mRNA | AY039011.1 | - | 2 - 2 | 549 | 89 |
BC125137.1 | - | 1 - 1 | 305 | 89 |