The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005707.1 | NP_005698.1 | 1 - 1 | 760 | 163 |
GenBank mRNA | AK096970.1 | - | 1 - 1 | 760 | 163 |
AK225816.1 | - | 1 - 1 | 727 | 163 | |
AK303203.1 | - | 2 - 2 | 500 | 163 |