The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145160.1 | NP_001138632.1 | 9 - 9 | 2169 | 125 |
NM_003290.2 | NP_003281.1 | 8 - 8 | 2184 | 125 | |
XM_011528231.2 | XP_011526533.1 | 9 - 9 | 2082 | 125 | |
XM_011528232.1 | XP_011526534.1 | 8 - 8 | 3402 | 125 | |
XM_017027188.1 | XP_016882677.1 | 9 - 9 | 2200 | 125 | |
XM_017027189.1 | XP_016882678.1 | 8 - 8 | 4035 | 125 | |
GenBank mRNA | FJ224305.1 | - | 8 - 8 | 1993 | 125 |
X05276.1 | - | 8 - 8 | 1993 | 125 |