The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037553.1 | NP_001032642.1 | 9 - 9 | 5905 | 73 |
NM_020132.4 | NP_064517.1 | 10 - 10 | 6025 | 73 | |
XM_005261160.4 | XP_005261217.1 | 10 - 10 | 6026 | 73 | |
XM_006724029.3 | XP_006724092.1 | 9 - 9 | 5954 | 73 | |
XM_006724030.3 | XP_006724093.1 | 9 - 9 | 5922 | 73 | |
XM_006724031.3 | XP_006724094.1 | 9 - 9 | 5925 | 73 | |
XM_011529660.2 | XP_011527962.1 | 9 - 9 | 5764 | 73 | |
XM_011529661.2 | XP_011527963.1 | 9 - 9 | 6079 | 73 | |
XM_011529662.2 | XP_011527964.1 | 11 - 11 | 6729 | 73 | |
XM_011529663.2 | XP_011527965.1 | 9 - 9 | 5938 | 73 | |
XM_011529664.2 | XP_011527966.1 | 9 - 9 | 5879 | 73 | |
XM_011529665.2 | XP_011527967.1 | 11 - 11 | 6380 | 73 | |
XM_017028408.1 | XP_016883897.1 | 9 - 9 | 5908 | 73 | |
XM_017028409.1 | XP_016883898.1 | 9 - 9 | 5986 | 73 | |
XM_017028410.1 | XP_016883899.1 | 9 - 9 | 5991 | 73 | |
XM_017028411.1 | XP_016883900.1 | 9 - 9 | 5864 | 73 | |
XM_017028412.1 | XP_016883901.1 | 10 - 10 | 6957 | 73 | |
XM_017028413.1 | XP_016883902.1 | 10 - 10 | 5925 | 73 | |
GenBank mRNA | AK074300.1 | - | 1 - 1 | 1948 | 73 |