The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316968.1 | NP_001303897.1 | 3 - 3 | 3114 | 102 |
NM_001316969.1 | NP_001303898.1 | 3 - 3 | 2830 | 102 | |
NM_033211.3 | NP_149988.1 | 3 - 3 | 2874 | 102 | |
GenBank mRNA | AK058020.1 | - | 3 - 3 | 2809 | 102 |
BC009203.2 | - | 3 - 3 | 2835 | 102 |