Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VRTN
Entrez Gene ID
55237
Gene Name
vertebrae development associated
Gene Aliases
C14orf115, vertnin
Chromosome Location
Chr.14: 74303076 - 74360008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018228.2 NP_060698.2 2 - 2 3362 104
GenBank mRNA AK001673.1 - 2 - 2 3316 104
BC053325.1 - 2 - 2 3345 104

View Full Product Details