Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- UBC
- Entrez Gene ID
-
7316
- Gene Name
- ubiquitin C
- Gene Aliases
-
HMG20
- Chromosome Location
-
Chr.12: 124911645 - 124915041 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M26880.1 |
- |
- |
2173 |
135 |
View Full Product Details