Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECEL1P2
Entrez Gene ID
347694
Gene Name
endothelin converting enzyme like 1 pseudogene 2
Gene Aliases
ECEL2
Chromosome Location
Chr.2: 232385750 - 232387044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028501.1 1 - 1 671 157

View Full Product Details