Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEUROG3
Entrez Gene ID
50674
Gene Name
neurogenin 3
Gene Aliases
Atoh5, Math4B, NGN-3, bHLHa7, ngn3
Chromosome Location
Chr.10: 69571440 - 69573454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020999.3 NP_066279.2 2 - 2 1137 127
XM_017016280.1 XP_016871769.1 2 - 2 1174 127
GenBank mRNA BC069098.1 - 2 - 2 1049 127

View Full Product Details