Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR13C9
Entrez Gene ID
286362
Gene Name
olfactory receptor family 13 subfamily C member 9
Gene Aliases
OR37L, OR9-13
Chromosome Location
Chr.9: 104617248 - 104618204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001956.1 NP_001001956.1 1 - 1 443 81

View Full Product Details