Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARMCX2
- Entrez Gene ID
-
9823
- Gene Name
- armadillo repeat containing, X-linked 2
- Gene Aliases
-
ALEX2, GASP9
- Chromosome Location
-
Chr.X: 101655281 - 101659891 on Build GRCh38
View Full Product Details