The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006683.4 | NP_006674.2 | 2 - 2 | 760 | 121 |
XM_017020934.1 | XP_016876423.1 | 2 - 2 | 777 | 121 | |
GenBank mRNA | AA769825.1 | - | 1 - 1 | 121 |