Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHF2
Entrez Gene ID
79666
Gene Name
pleckstrin homology and FYVE domain containing 2
Gene Aliases
EAPF, PHAFIN2, ZFYVE18
Chromosome Location
Chr.8: 95133721 - 95156685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024613.3 NP_078889.1 2 - 2 2538 147
GenBank mRNA AK023249.1 - 2 - 2 2311 147
AL834473.1 - 2 - 2 2461 147

View Full Product Details