Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SMR3B
- Entrez Gene ID
-
10879
- Gene Name
- submaxillary gland androgen regulated protein 3B
- Gene Aliases
-
P-B, PBII, PRL3, PROL3, SMR1B
- Chromosome Location
-
Chr.4: 70383078 - 70390244 on Build GRCh38
- Gene Symbol
- SMR3A
- Entrez Gene ID
-
26952
- Gene Name
- submaxillary gland androgen regulated protein 3A
- Gene Aliases
-
P-B1, PBI, PRL5, PROL5
- Chromosome Location
-
Chr.4: 70360776 - 70367158 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CD722466.1 |
- |
4 - 4 |
391 |
132 |
View Full Product Details