The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_038866.1 | 3 - 3 | 2544 | 107 | |
GenBank mRNA | AK096387.1 | - | 3 - 3 | 2544 | 107 |