Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAIP2
Entrez Gene ID
51247
Gene Name
poly(A) binding protein interacting protein 2
Gene Aliases
PAIP-2, PAIP2A
Chromosome Location
Chr.5: 139341832 - 139369720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017009522.1 XP_016865011.1 1 - 1 235 74

View Full Product Details