Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
Chromosome Location
Chr.Y: 18872501 - 19077547 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF119903.1 - LIT2233
CYorf14
1 - 1 421 104


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI207481.1 - 1 421 Chr. Y - 18877477 - 18878228 [-] on Build GRCh38 104
GenBankmRNA AA662971.1 - 1 Chr. Y - 18877471 - 18877924 [+] on Build GRCh38 104


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream heat shock transcription factor, Y-linked 2 HSFY2 HSF2L, HSFY -103.7kb Anti-sense Chr. - 18731440 - 18790733 on Build GRCh38
Downstream BCL6 corepressor pseudogene 1 BCORP1 BCORL2 577.5kb Anti-sense Chr. - 19455431 - 19503153 on Build GRCh38

View Full Product Details