Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
Chromosome Location
Chr.1: 12166948 - 12209222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001066.2 NP_001057.1 10 - 10 3626 120
XM_011542060.2 XP_011540362.1 11 - 11 3750 120
XM_011542063.2 XP_011540365.1 10 - 10 3684 120
XM_017002214.1 XP_016857703.1 10 - 10 3650 120
XM_017002215.1 XP_016857704.1 9 - 9 3581 120
GenBank mRNA BC052977.1 - 10 - 10 3626 120
M32315.1 - 10 - 10 3627 120
S63368.1 - 9 - 9 3428 120

View Full Product Details