The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011525489.2 | XP_011523791.1 | 9 - 9 | 4662 | 112 |
XM_017025357.1 | XP_016880846.1 | 9 - 9 | 4735 | 112 | |
XM_017025358.1 | XP_016880847.1 | 10 - 10 | 4197 | 112 | |
XM_017025359.1 | XP_016880848.1 | 9 - 9 | 4048 | 112 | |
XM_017025360.1 | XP_016880849.1 | 9 - 9 | 4098 | 112 | |
XM_017025361.1 | XP_016880850.1 | 9 - 9 | 4354 | 112 | |
XM_017025362.1 | XP_016880851.1 | 10 - 10 | 3816 | 112 | |
XM_017025364.1 | XP_016880853.1 | 8 - 8 | 4256 | 112 |