Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK094652.1 |
- |
1 |
2148 |
|
Chr. 8 - 56067252 - 56071789 [-] on Build GRCh38 |
107 |
GenBankmRNA |
DB302111.1 |
- |
2 |
|
|
Chr. 8 - 56067252 - 56069996 [+] on Build GRCh38 |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ribosomal protein S20 |
RPS20 |
S20 |
|
Anti-sense |
Chr. - 56068180 - 56074581 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 54 |
SNORD54 |
RNU54, U54 |
3.8kb
|
Anti-sense |
Chr. - 56073839 - 56073901 on Build GRCh38 |
View Full Product Details