Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
MAN2B1 | NM_000528.3 | NP_000519.2 | ||
NM_001173498.1 | NP_001166969.1 | |||
XM_005259913.1 | XP_005259970.1 | |||
XM_017026818.1 | XP_016882307.1 | |||
AB209921.1 | BAD93158.1 | |||
AK291572.1 | ||||
AK303418.1 | ||||
AW172353.1 | ||||
BC000736.2 | AAH00736.1 | |||
U05572.1 | AAB03816.1 | |||
U60266.1 | AAC34130.1 | |||
U68382.1 | AAC50811.1 | |||
U68567.1 | AAC50812.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160586 | Chr19:12597480 - 12823909 on Build GRCh38 | Duplication | RNASEH2A WDR83 C19orf43 SNORD41 SNORD135 ZNF791 FBXW9 DHPS LOC105372280 PRDX2 TNPO2 MAN2B1 ASNA1 JUNB BEST2 WDR83OS HOOK2 MIR5684 ZNF490 |
dgv138n111 | Chr19:12627853 - 12810372 on Build GRCh38 | Deletion | RNASEH2A WDR83 C19orf43 SNORD41 SNORD135 ZNF791 FBXW9 DHPS LOC105372280 PRDX2 TNPO2 MAN2B1 ASNA1 JUNB BEST2 WDR83OS HOOK2 MIR5684 |