Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
Chromosome Location
Chr.10: 13643706 - 14330952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_134578.1 4 - 4 1136 149

View Full Product Details