The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011537951.2 | XP_011536253.1 | 1 - 1 | 2035 | 75 |
XM_011537952.2 | XP_011536254.1 | 1 - 1 | 2035 | 75 | |
XM_017018851.1 | XP_016874340.1 | 1 - 1 | 121 | 75 |