Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRR
Entrez Gene ID
63826
Gene Name
serine racemase
Gene Aliases
ILV1, ISO1
Chromosome Location
Chr.17: 2303371 - 2325267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721566.3 XP_006721629.1 1 - 1 786 101

View Full Product Details