Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OPN1MW2
- Entrez Gene ID
-
728458
- Gene Name
- opsin 1 (cone pigments), medium-wave-sensitive 2
- Gene Aliases
-
GOP
- Chromosome Location
-
Chr.X: 154219734 - 154233286 on Build GRCh38
- Gene Symbol
- OPN1LW
- Entrez Gene ID
-
5956
- Gene Name
- opsin 1 (cone pigments), long-wave-sensitive
- Gene Aliases
-
CBBM, CBP, COD5, RCP, ROP
- Chromosome Location
-
Chr.X: 154144224 - 154159032 on Build GRCh38
- Gene Symbol
- OPN1MW
- Entrez Gene ID
-
2652
- Gene Name
- opsin 1 (cone pigments), medium-wave-sensitive
- Gene Aliases
-
CBBM, CBD, COD5, GCP, GOP, OPN1MW1
- Chromosome Location
-
Chr.X: 154182596 - 154196861 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_000513.2 |
NP_000504.1 |
6 - 6 |
1171 |
95 |
GenBank mRNA |
BC140938.1 |
- |
6 - 6 |
1171 |
95 |
- Gene Symbol
- OPN1MW3
- Entrez Gene ID
-
101060233
- Gene Name
- opsin 1 (cone pigments), medium-wave-sensitive 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 154257620 - 154271068 on Build GRCh38
View Full Product Details