The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003268.5 | NP_003259.2 | 6 - 6 | 3227 | 89 |
XM_005273241.4 | XP_005273298.2 | 6 - 6 | 3173 | 89 | |
XM_005273242.4 | XP_005273299.2 | 7 - 7 | 3293 | 89 | |
XM_005273243.4 | XP_005273300.2 | 5 - 5 | 3079 | 89 | |
XM_006711504.3 | XP_006711567.1 | 5 - 5 | 3118 | 89 | |
XM_006711505.3 | XP_006711568.1 | 4 - 4 | 3024 | 89 | |
XM_006711506.3 | XP_006711569.1 | 3 - 3 | 2826 | 89 | |
XM_011509937.2 | XP_011508239.1 | 6 - 6 | 3234 | 89 | |
XM_017002208.1 | XP_016857697.1 | 5 - 5 | 3140 | 89 | |
GenBank mRNA | AB060695.1 | - | 6 - 6 | 3289 | 89 |
AF051151.1 | - | 6 - 6 | 3289 | 89 | |
BC109118.1 | - | 4 - 4 | 2924 | 89 | |
BC109119.1 | - | 4 - 4 | 2924 | 89 | |
U88881.1 | - | 1 - 1 | 1107 | 89 |