The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003469.4 | NP_003460.2 | 2 - 2 | 2205 | 116 |
GenBank mRNA | AK298828.1 | - | 3 - 3 | 1646 | 116 |
BC022509.1 | - | 2 - 2 | 2113 | 116 | |
M25756.1 | - | 2 - 2 | 2038 | 116 |