Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB6
Entrez Gene ID
10773
Gene Name
zinc finger and BTB domain containing 6
Gene Aliases
ZID, ZNF482
Chromosome Location
Chr.9: 122908056 - 122913328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006626.5 NP_006617.1 2 - 2 4037 150
GenBank mRNA AK098650.1 - 1 - 1 1469 150
BC037282.2 - 2 - 2 4039 150

View Full Product Details