The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006794.3 | NP_006785.1 | 2 - 2 | 2062 | 84 |
GenBank mRNA | AF072693.1 | - | 2 - 2 | 1957 | 84 |
BC067475.1 | - | 1 - 1 | 1858 | 84 |