Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5F1
Entrez Gene ID
338674
Gene Name
olfactory receptor family 5 subfamily F member 1
Gene Aliases
OR11-10
Chromosome Location
Chr.11: 55993681 - 55994625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003697.1 NP_003688.1 1 - 1 542 115
GenBank mRNA BC101239.2 - 1 - 1 544 115
BC101241.1 - 1 - 1 542 115
BC101242.3 - 1 - 1 541 115

View Full Product Details